missing translation for 'onlineSavingsMsg'
Learn More

Melford Agarose ME

Product Code. 30017980
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30017980 500 g 500g
30017979 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30017980 Supplier Melford Supplier No. A20085500.0
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For DNA/RNA fragments >1000bp.

For DNA/RNA fragments >1000bp. Intermediate EEO.

Specifications

Chemical Name or Material Agarose ME
CAS 9012-36-6
Quantity 500 g
Grade Molecular Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.