missing translation for 'onlineSavingsMsg'
Learn More

Melford Agarose Low Melt

Product Code. 30017972
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
Unit Size:
100g
250g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30017972 500 g 500g
30017971 250 g 250g
30017970 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30017972 Supplier Melford Supplier No. A20070500.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ideal gelling properties for use in cloning assays or for recovery of heat labile samples following electrophoresis.

Ideal gelling properties for use in cloning assays or for recovery of heat labile samples following electrophoresis.

Gel Strength, 1%: >200 g/cm2

Gelling Temperature: 26°C to 30°C

Melting Temperature: <65°C

Sulfate: <0.10%

EEO: <0.10

DNase and RNase: None Detected

Specifications

Chemical Name or Material Agarose, Low Melt
CAS 9012-36-6
Quantity 500 g
Grade Molecular Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.