missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Agarose LM, low melt, for recovery of samples after separation

Product Code. 11401380
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11401380 25 g 25g
11491370 5 g 5g
11411380 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11401380 Supplier Thermo Scientific Chemicals Supplier No. H26417.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for size-based separation of nucleic acids in agarose gel electrophoresis applications. | CAS: 9012-36-6 | C24H38O19 | 630.55 g/mol

Used for size-based separation of nucleic acids in agarose gel electrophoresis applications. Ideal for analysis and recovery of DNA and RNA for routine applications. Ideal for a variety of applications such as in-gel manipulation, tissue and cell culture, and viral plaque assays. Can also be used in protein electrophoresis applications such as Ouchterlony (antigen-antibody interaction assay) and radial immunodiffusion (RID) (antigen quantitation assay)

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9012-36-6

Specifications

Molecular Formula C24H38O19
InChI Key MJQHZNBUODTQTK-WKGBVCLCSA-N
SMILES OC[C@H]1O[C@@H](O[C@@H]2[C@@H]3CO[C@H]2[C@H](O)[C@H](O[C@H]2[C@@H](O)[C@@H](CO)O[C@@H](O[C@@H]4[C@@H]5CO[C@H]4[C@H](O)[C@H](O)O5)[C@@H]2O)O3)[C@H](O)[C@@H](O)[C@H]1O
IUPAC Name (2S,3R,4S,5R,6R)-2-{[(1S,3S,4S,5S,8R)-3-{[(2S,3R,4S,5S,6R)-2-{[(1S,3R,4S,5S,8R)-3,4-dihydroxy-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy}-3,5-dihydroxy-6-(hydroxymethyl)oxan-4-yl]oxy}-4-hydroxy-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Weight (g/mol) 630.55
Quantity 25 g
Sensitivity Hygroscopic; Moisture sensitive
Product Type LM Agarose

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.