missing translation for 'onlineSavingsMsg'
Learn More

Agarose LE, Molecular Biology Grade, Ultrapure

Product Code. 15855358 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
Packaging:
Plastic Bottle
Unit Size:
100g
1kg
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15855358 1 kg Plastic Bottle 1kg
15825208 25 g Plastic Bottle 25g
15835208 100 g Plastic Bottle 100g
15855208 500 g Plastic Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15855358 Supplier Thermo Scientific Supplier No. J32802.A1
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 9012-36-6

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9012-36-6
MDL Number MFCD00081294

Specifications

Physical Form Granular Free-flowing Powder
Quantity 1 kg
Packaging Plastic Bottle
Grade Molecular Biology
Chemical Name or Material Agarose LE
What are the applications for Agarose LE, Molecular Biology Grade, Ultrapure?

Applications:
1. Routine separation of DNA or RNA fragments.
2. PCR analysis.
3. Blotting applications such as southern blots and northern blots.
4. Protein analysis such as immunodiffusion, rocket assays and immuno electrophoresis.

What are the separation guidelines for Agarose LE, Molecular Biology Grade, Ultrapure?

Range of separation in 1X TAE: 0.6%: 20,000-1000 bp
0.8%: 12,000-500 bp
1.0%: 8000-300 bp
1.2%: 6000-200 bp
1.5%: 3500-100 bp
2.0%: 2000-50 bp

Range of separation in 1X TBE:
0.6%: 15,000-1000 bp
0.8%: 10,000-500 bp
1.0%: 7000-250 bp
1.2%: 5000-200 bp
1.5%: 3000-100 bp
2.0%: 2000-50 bp

How do I prepare an agarose gel?

We recommend the following to prepare an agarose gel:

  1. Weigh out the desired amount of agarose and place it in an Erlenmeyer flask with a measured amount of electrophoresis buffer. Make sure to use the same electrophoresis buffer in the gel as for the running buffer.
  2. Example: For a 0.8% gel, add 0.8 g of agarose and 100 mL of TBE Buffer (1X), to a 200 mL flask. The larger flask insures against the agarose boiling over.
  3. Dissolve the agarose in a boiling water bath or in a revolving-plate microwave oven. All the grains of agarose should be dissolved, and the solution should be clear.
  4. Cool the solution to 60°C (70°C for concentrations 2% or above) and pour immediately.
  5. Allow the gel to set for 30 minutes before using.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.