missing translation for 'onlineSavingsMsg'
Learn More

Agarose High Resolution, MP Biomedicals™

Product Code. p-7139917 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11456960 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11456960 Supplier MP Biomedicals Supplier No. 11AGAR0050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Agarose High Resolution, Molecular Biology Grade, White Powder

  • Agarose high resolution is an intermediate melting and gelling point agarose which exhibits superior sieving characteristics.
  • It is recommended for analytical gels of DNA fragments under 1000 bp and is ideal for small DNA fragments down to 20 bp can be separated on 6 % gels.
  • It resolves fragments differing in size by only 2 bp.
  • Its use avoids the need for PAGE Southern and Northern transfers can be achieved efficiently from them at concentrations up to 2%.
  • DNA fragments can be purified with high efficiency from this agarose gels up to 4% by using either a purification kit or an electroelution system
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9012-36-6
Molecular Formula C24H38O19
Molecular Weight (g/mol) 630.55
MDL Number MFCD00081294
InChI Key MJQHZNBUODTQTK-WKGBVCLCSA-N
Synonym Galactan Hydrocolloid, 3,6-Anhydro-α-L-galacto-β-D-galactan, FastLane agarose, Indubiose A4

Specifications

Physical Form Powder
Melting Point < 67°C (1.5%gel)
Color White
Quantity 50 g
Electroendosmosis (EEO) ≤0.12
Gelation Temperature < 26°C (1.5%gel)
Gel Strength > 600g/cm2 (1.5%gel)
Solubility Information Soluble in water (15mg/mL).
Grade Molecular Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.