missing translation for 'onlineSavingsMsg'
Learn More

Agarose, Electrophoresis Grade, MP Biomedicals™

Product Code. 11462940 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
50 g
250 g
500 g
Unit Size:
10g
250g
25g
500g
50g
5g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11462940 500 g 500g
11402940 5 g 5g
11412940 10 g 10g
11422940 25 g 25g
11432940 50 g 50g
11452940 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 11462940 Supplier MP Biomedicals Supplier No. 0210026790
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 9012-36-6
MDL Number MFCD00081294
Synonym 3, 6-Anhydro-α-L-galacto-β-D-galactan, FastLane agarose

Specifications

Physical Form Powder
Density 1.00g/mL at 20°C
Color White
Quantity 500 g
Grade Electrophoresis
Chemical Name or Material Agarose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.