missing translation for 'onlineSavingsMsg'
Learn More

Melford Agarose 500 mg Tablets

Product Code. 30017986
Click to view available options
Quantity:
100 g, 200 Tablets
50 g, 100 Tablets
Unit Size:
100 tablets
200 tablets
This item is not returnable. View return policy

Product Code. 30017986

Brand: Melford A2009650.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Save time and labor preparing agarose gels for electrophoresis of nucleic acids.

Save time and labor preparing agarose gels for electrophoresis of nucleic acids. Just place the required number of tablets into a standard buffer, then mix and heat until completely dissolved.

Tablet Weight Range: 485 to 515 mg

Gel Strength, 1.5%: >1200 g/cm2

Gelling Range: 36°C to 39°C

Melting Temp: 87°C to 89°C

EEO: <0.10

Sulfate: <0.15

DNase, RNase: None Detected

Specifications

Chemical Name or Material Agarose, Tablets
CAS 9012-36-6
Quantity 50 g, 100 Tablets
Grade Molecular Biology
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.