missing translation for 'onlineSavingsMsg'
Learn More

Melford Agarose

Product Code. 30017983
Click to view available options
Quantity:
50 g
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
50g
This item is not returnable. View return policy

Product Code. 30017983

Brand: Melford A2009050.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Molecular Biology Grade Agarose.

Molecular Biology Grade Agarose. High Purity for preparative and analytical electrophoresis of nucleic acids ≥1000bp. Forms a solid gel at low concentration. Product is RNase/DNase free.

Gel Strength, 1%: ≥1200 g/cm2

Gel Strength, 1.5%: ≥2700 g/cm2

Gel Point: 35°C to 37°C

Melting Point: 87°C to 89°C

EEO: 0.09 to 0.12

Sulfate: ≥0.03%

Loss on Drying: <10%

DNA Binding: None Detected

Specifications

Chemical Name or Material Agarose
CAS 9012-36-6
Quantity 50 g
Grade Molecular Biology
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.