missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Agar Technical No.2

Product Code. 15870542
Change view
Click to view available options
Quantity:
500 g
5 kg
Unit Size:
500g
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15870542 5 kg 5kg
15562141 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15870542 Supplier Thermo Scientific™ Supplier No. LP0012T

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Oxoid Agar Technical (Agar No. 2) is a general-purpose bacteriological-grade agar.

A technical grade agar which conforms to the specifications of the U.S. Pharmacopoeia and A.P.H.A. Suitable for applications where clarity and compatibility are not of prime importance. It is of high gel strength and will give a firm gel 1.2% w/v.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Agar
Quantity 5 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.