missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Agar, NF Grade
Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
Unit Size:
100g
1kg
250g
500g
Description
- 80-100 mesh
- Substitute for gelatin, isinglass
- Used as nutrient media for bacterial culture
Specifications
Specifications
| Content And Storage | 15°C to 30°C |
| Form | Powder |
| Grade | USP, NF |
| Product Type | Agar |
| For Use With (Application) | Used for nutrient media for bacterial cultures |
| Quantity | 500 g |
Safety and Handling
Signal Word
- Danger
- Flammable solid Category 1
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H228-Flammable solid.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Agar
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction