missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Agar, NF Grade
Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
Unit Size:
100g
1kg
250g
500g
Description
- 80-100 mesh
- Substitute for gelatin, isinglass
- Used as nutrient media for bacterial culture
Specifications
Specifications
| Content And Storage | 15°C to 30°C |
| Form | Powder |
| Grade | USP, NF |
| Product Type | Agar |
| For Use With (Application) | Used for nutrient media for bacterial cultures |
| Quantity | 250 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction