missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford Agar Capsules
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
Description
Each capsule contains 1g Agar.
Each capsule contains 1g Agar. Convenient and easy to use, simply add required number of premeasured capsules according to formulation being prepared without weighing or measuring.
Specifications
Specifications
| Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction