Learn More
Agar (80-100 Mesh), Fisher BioReagents
Brand: Fisher Bioreagents BP2641-100
84.62 GBP valid until 2024-12-31
Use promo code "23286" to get your promotional price.
Code : 32
Additional Details : CAS Number : 9002-18-0 Weight : 0.15000kg
Specifications
Solid | |
Agar | |
Poly Bottle | |
100 g |