missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AGAP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562401
This item is not returnable.
View return policy
Description
Immunogen sequence: PDERFVLMAT PNLSRRNNNR NNSSGRVPTI I Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 84%.
GTPase-activating protein for ARF1 and, to a lesser extent, ARF5. Directly and specifically regulates the adapter protein 3 (AP-3)-dependent trafficking of proteins in the endosomal-lysosomal system.
Specifications
| AGAP1 | |
| Polyclonal | |
| Unconjugated | |
| AGAP1 | |
| AGAP1; AGAP-1; Arf GAP with GTP-binding protein-like, ANK repeat and PH domains 1; ArfGAP with GTPase domain, ankyrin repeat and PH domain 1; arf-GAP with GTPase, ANK repeat and PH domain-containing protein 1; centaurin, gamma 2; centaurin-gamma-2; CENTG2; cnt-g2; GGAP1; GTP-binding and GTPase-activating protein 1; Kiaa1099; mKIAA1099; R75364 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 116987 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UPQ3 | |
| AGAP1 | |
| Recombinant protein corresponding to Human CENTG2. Recombinant protein control fragment (Product #RP-101393). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction