missing translation for 'onlineSavingsMsg'
Learn More

AGA Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£190.00 - £255.00

Specifications

Antigen AGA
Dilution ELISA 1:5000-1:10000, Immunohistochemistry-Paraffin 1:500-1:2000
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18680026
View Documents
Novus Biologicals
NBP2-99967-100ul
100 μg
£255.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18688455
View Documents
Novus Biologicals
NBP2-99967-50ul
50 μg
£190.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

AGA Polyclonal antibody specifically detects AGA in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
Specifications

Specifications

AGA
ELISA, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Cell Cycle and Replication, Proteases & Other Enzymes
0.2 um filtered solution in PBS
175
IgG
Antigen and protein A Affinity-purified
ELISA 1:5000-1:10000, Immunohistochemistry-Paraffin 1:500-1:2000
Polyclonal
Purified
RUO
Human
AGU, aspartylglucosaminidase, aspartylglucosylamine deaspartylase, ASRG, EC 3.5.1, EC 3.5.1.26, GA, glycosylasparaginase, N(4)-(beta-N-acetylglucosaminyl)-L-asparaginase, N4-(N-acetyl-beta-glucosaminyl)-L-asparagine amidase
Produced in rabbits immunized with purified, recombinant Human AGA (Accession#: CAA39029.1; Met1-Ile346).
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.