missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AG1024, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Description
AG1024 (Tyrphostin AG 1024) is a reversible, competitive and selective IGF-1R inhibitor with an IC50 of 7 μM. AG1024 inhibits phosphorylation of IR (IC50=57 μM). AG1024 induces apoptosis and has anti-cancer activity.
Specifications
Specifications
| Chemical Name or Material | AG1024 |
| CAS | 65678-07-1 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C14H13BrN2O |
| Quantity | 5 mg |
| Synonym | Tyrphostin AG 1024 |
| Solubility Information | DMSO : ≥ 50 mg/mL (163.84 mM) |
| SMILES | N#C/C(C#N)=C\C1=CC(C(C)(C)C)=C(O)C(Br)=C1 |
| Molecular Weight (g/mol) | 305.17 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction