Learn More
LNI Swissgas AG OFCAS iQ 220 8
Product Code. 17882039 Shop All LNI Swissgas Products
Change view
Click to view available options
Voltage:
100/115/220/230 V
Unit Size:
Each
Product Code. Voltage unitSize
17882039 100/115/220/230 V Each
1 options
Product Code. 17882039 Supplier LNI Swissgas Supplier No. 6920.47.220
  • ONLINE EXCLUSIVE PRICE
    £11955.00
    £11743.48 / Pack of 1
    Save £211.52
    1% Off
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
AG OFCAS-Scroll with dryer integrated.
  • AG OFCAS-Scroll 220 L/min at 8 bar, 230 V with dryer integrated.
Specifications
Voltage 100/115/220/230 V
Type Air Generator