missing translation for 'onlineSavingsMsg'
Learn More

Xylem AG Multi Refractometer Calibration Fluid 5 x 5 mL (UKAS/NIST)

Product Code. 30484479
Change view
Click to view available options
For Use With (Application):
Beverage (USA)
Juice
Petrochemical
Pharma and Chem
Soda
Unit Size:
25mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) Unit Size
30484479 Petrochemical 25mL
30484474 Soda 25mL
30484454 Beverage (USA) 25mL
30484476 Juice 25mL
30484471 Pharma and Chem 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 30484479 Supplier Xylem Supplier No. 90914
  • ONLINE EXCLUSIVE PRICE
    £154.00
    £150.28 / 25mL
    Save £3.72
    2% Off
View all promotions
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Calibration fluid for calibration of refractometers.

  • Calibration Fluid
  • For Refractometers
TRUSTED_SUSTAINABILITY

Specifications

Type Calibration Fluid
Refractive Index Mixed
For Use With (Equipment) Refractometer
For Use With (Application) Petrochemical
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.