missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ag, Al, B, Ba, Bi, Ca, Cd, Cr, Cu, Fe, In, Li, K, Mg, Mn, Mo, Na, Ni, P, Pb, Sb, Si, Sn, Ti, V, Zn @ 100 μg/g in 75 cSt Hydrocarbon Oil, VHG™
Click to view available options
Quantity:
100 g
200 g
400 g
Unit Size:
100g
200g
400g
Description
Ag, Al, B, Ba, Bi, Ca, Cd, Cr, Cu, Fe, In, Li, K, Mg, Mn, Mo, Na, Ni, P, Pb, Sb, Si, Sn, Ti, V, Zn @ 100 μg/g in 75 cSt Hydrocarbon Oil
Specifications
Specifications
| Chemical Name or Material | Aluminium/Antimony/Barium/Bismuth/Boron/Cadmium/Calcium/Chromium/Copper/Indium/Iron/Lead/Lithium/Magnesium/Manganese/Molybdenum/Nickel/Phosphorus/Potassium/Silicon/Silver/Sodium/Tin/Titanium/Vanadium/Zinc |
| CAS | 7429-90-5, 7440-36-0, 7440-39-3, 7440-69-9, 7440-42-8, 7440-43-9 |
| Methods | ICP/XRF/RDE |
| Analyte or Component Names | Aluminium/Antimony/Barium/Bismuth/Boron/Cadmium/Calcium/Chromium/Copper/Indium/Iron/Lead/Lithium/Magnesium/Manganese/Molybdenum/Nickel/Phosphorus/Potassium/Silicon/Silver/Sodium/Tin/Titanium/Vanadium/Zinc |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Hydrocarbon Oil |
| Quantity | 400 g |
| Concentration or Composition (by Analyte or Components) | Aluminium 100 μg/g/Antimony 100 μg/g/Barium 100 μg/g/Bismuth 100 μg/g/Boron 100 μg/g/Cadmium 100 μg/g/Calcium 100 μg/g/Chromium 100 μg/g/Copper 100 μg/g/Indium 100 μg/g/Iron 100 μg/g/Lead 100 μg/g/Lithium 100 μg/g/Magnesium 100 μg/g/Manganese 100 μg/g/Molybdenum 100 μg/g/Nickel 100 μg/g/Phosphorus 100 μg/g/Potassium 100 μg/g/Silicon 100 μg/g/Silver 100 μg/g/Sodium 100 μg/g/Tin 100 μg/g/Titanium 100 μg/g/Vanadium 100 μg/g/Zinc 100 μg/g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction