missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AG-3/AGR3 Antibody (AGR3.4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-62196
This item is not returnable.
View return policy
Description
AG-3/AGR3 Monoclonal antibody specifically detects AG-3/AGR3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| AG-3/AGR3 | |
| Monoclonal | |
| Unconjugated | |
| PBS and 0.2% (w/v) high grade BSA (protease free) | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| AGR3.4 | |
| Western Blot 1 μg/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/ Immunofluorescence 1 μg/ml, Immunohistochemistry-Paraffin 5 μg/ml | |
| AG-3, anterior gradient homolog 3 (Xenopus laevis), BCMP11AG3, Breast cancer membrane protein 11, hAG-3member 18 | |
| Purified human AGR3 protein | |
| 0.1 mg | |
| Cancer | |
| 155465 | |
| Store at 4°C. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction