missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AG-2/AGR2 Antibody (10E2), mFluor Violet 500 SE, Novus Biologicals™
Description
AG-2/AGR2 Monoclonal antibody specifically detects AG-2/AGR2 in Human, Mouse, Bovine, Canine, Equine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | AG-2/AGR2 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 10E2 |
| Conjugate | mFluor Violet 500 SE |
| Dilution | Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate |
| Gene Alias | AG-2, AG2member 17, anterior gradient 2 homolog (Xenopus laevis), anterior gradient homolog 2 (Xenopus laevis), GOB-4, hAG-2, HPC8, secreted cement gland homolog |
| Host Species | Mouse |
| Immunogen | A portion of amino acids 50-100 of human AGR2 was used as the immunogen for the antibody. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?