missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AG-2/AGR2 Antibody (10E2) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-80564
This item is not returnable.
View return policy
Description
AG-2/AGR2 Monoclonal specifically detects AG-2/AGR2 in Human, Mouse, Bovine, Canine, Equine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| AG-2/AGR2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| Western Blot : 4-6 μg/mL, Immunohistochemistry 5 μg/mL, Immunohistochemistry-Paraffin | |
| AG-2, AG2member 17, anterior gradient 2 homolog (Xenopus laevis), anterior gradient homolog 2 (Xenopus laevis), GOB-4, hAG-2, HPC8, secreted cement gland homolog | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Orangutan, Panda and Platypus. Immunogen sequence similarity with other species: Xenopus (82%), Chicken (83%). | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1000 | |
| Unconjugated | |
| PBS | |
| AGR2 | |
| A portion of amino acids 50-100 of human AGR2 was used as the immunogen for the antibody. | |
| 0.1 mg | |
| Breast Cancer, Cancer, Stem Cell Markers | |
| 10551 | |
| Human, Mouse, Bovine, Canine, Equine, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction