missing translation for 'onlineSavingsMsg'
Learn More

AG-1478, MedChemExpress

Product Code. 16475728
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16475728

Brand: Medchem Express HY13524

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AG-1478 (Tyrphostin AG-1478) is a selective EGFR tyrosine kinase inhibitor with IC50 of 3 nM. AG-1478 has antiviral effects against HCV and encephalomyocarditis virus (EMCV).

AG-1478 (Tyrphostin AG-1478) is a selective EGFR tyrosine kinase inhibitor with IC50 of 3 nM. AG-1478 has antiviral effects against HCV and encephalomyocarditis virus (EMCV).

Specifications

Chemical Name or Material AG-1478
Physical Form Solid
CAS 153436-53-4
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C16H14ClN3O2
For Use With (Application) Cancer-Kinase/protease
Synonym Tyrphostin AG-1478 NSC 693255
Solubility Information DMSO : 50 mg/mL (158.35 mM; Need ultrasonic)
SMILES COC1=CC2=NC=NC(NC3=CC=CC(Cl)=C3)=C2C=C1OC
Molecular Weight (g/mol) 315.75
Formula Weight 315.75
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.