missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Afuresertib hydrochloride, MedChemExpress
Description
Afuresertib hydrochloride (GSK 2110183 hydrochloride) is an orally bioavailable, selective, ATP-competitive and potent pan-Akt kinase inhibitor with Kis of 0.08/2/2.6 nM for Akt1/Akt2/Akt3 respectively.
Specifications
Specifications
| Chemical Name or Material | Afuresertib hydrochloride |
| CAS | 1047645-82-8 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C18H18Cl3FN4OS |
| Quantity | 100 mg |
| Synonym | GSK2110183 hydrochloride |
| Solubility Information | DMSO : 100 mg/mL (215.62 mM; Need ultrasonic) |
| SMILES | O=C(C1=CC(C2=C(Cl)C=NN2C)=C(Cl)S1)N[C@@H](CC3=CC=CC(F)=C3)CN.[H]Cl |
| Molecular Weight (g/mol) | 463.78 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?