missing translation for 'onlineSavingsMsg'
Learn More

AFP Monoclonal Antibody (F1-6P2A8-P2B9A9), Invitrogen™

Product Code. 12837952
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12837952 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12837952 Supplier Invitrogen Supplier No. MIA1301

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MIA1301 can be used for immunofluorescence analysis of AFP in the endoderm derived from human embryonic stem cells. By Western blot, MIA1301 detects endogenous AFP protein in the early hepatocye-like cells derived from human embryonic stem cells. MIA1301 was formerly sold as a Seradyn product.

AFP (Alpha Fetoprotein) is a major plasma protein produced by the yolk sac, intestinal tract and the liver during fetal life. AFP expression in adults is often associated with hepatoma or teratoma. However, hereditary persistence of AFP may also be found in individuals with no obvious pathology. The protein is thought to be the fetal counterpart of serum albumin, and AFP and albumin genes are present in tandem in the same transcriptional orientation on chromosome 4. AFP is found in monomeric, dimeric, and trimeric forms. AFP can also bind to copper ions, nickel ions, fatty acids and bilirubin. The level of AFP in amniotic fluid is used to measure renal loss of protein to screen for spina bifida and anencephaly. High level of serum AFP has been identified in patients with hepatocellular carcinomas (HCC), teratoblastoma, colorectal cancer, pancreatic cancer, and germ cell neoplasms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AFP
Applications ELISA, Immunocytochemistry, Immunoprecipitation, Radioimmune Assays (RIA), Western Blot
Classification Monoclonal
Clone F1-6P2A8-P2B9A9
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene AFP
Gene Accession No. P02771, P02772
Gene Alias a fetoglobulin; a fetoprotein; Afp; AFPD; Alpha fetoglobulin; alpha fetoprotein; alpha-1-fetoprotein; Alpha-fetoglobulin; alpha-fetoprotein; alpha-fetoprotein precursor; alpha-foetoprotein; chloride intracellular channel 6; FETA; fetoglobulin; fetoprotein; HPAFP; a fetoglobulin; a fetoprotein
Gene Symbols AFP
Host Species Mouse
Immunogen Purified AFP from Cord Blood
Purification Method Protein A
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11576, 174
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.