missing translation for 'onlineSavingsMsg'
Learn More

Afoxolaner 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30327405
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30327405 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30327405 Supplier Dr. Ehrenstorfer Supplier No. DREA10047600AL100
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Afoxolaner
CAS 1093861-60-9
Analyte or Component Names Afoxolaner
Shipping Condition +20°C
Molecular Formula C26 H17 Cl F9 N3 O3
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym 4-[5-[3-Chloro-5-(trifluoromethyl)phenyl]-4,5-dihydro-5-(trifluoromethyl)-3-isoxazolyl]-N-[2-oxo-2-[(2,2,2-trifluoroethyl)amino]ethyl]-1-naphthalenecarboxamide, 4-[5-(3-chloro-5-trifluoromethylphenyl)-5-trifluoromethyl-4,5-dihydro-isoxazol-3-yl]-naphthalene-1-carboxylic acid [(2,2,2-trifluoroethylcarbamoyl)-methyl]-amide, Afoxolaner, Nexgard
InChI Formula InChI=1S/C26H17ClF9N3O3/c27-15-8-13(7-14(9-15)25(31,32)33)23(26(34,35)36)10-20(39-42-23)18-5-6-19(17-4-2-1-3-16(17)18)22(41)37-11-21(40)38-12-24(28,29)30/h1-9H,10-12H2,(H,37,41)(H,38,40)
SMILES FC(F)(F)CNC(=O)CNC(=O)c1ccc(C2=NOC(C2)(c3cc(Cl)cc(c3)C(F)(F)F)C(F)(F)F)c4ccccc14
IUPAC Name 4-[5-[3-chloro-5-(trifluoromethyl)phenyl]-5-(trifluoromethyl)-4H-1,2-oxazol-3-yl]-N-[2-oxo-2-(2,2,2-trifluoroethylamino)ethyl]naphthalene-1-carboxamide
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 625.87
Quantity 1 mL
Formula Weight 625.0815 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.