missing translation for 'onlineSavingsMsg'
Learn More

AFN-1252, MedChemExpress

Product Code. 16449737
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16449737

Brand: Medchem Express HY16911

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AFN-1252(Debio 1452) is a potent inhibitor of enoyl-acyl carrier protein reductase (FabI), inhibited all clinical isolates of Staphylococcus aureus and Staphylococcus epidermidis at concentrations of ≤0.12 μg/ml.

AFN-1252(Debio 1452) is a potent inhibitor of enoyl-acyl carrier protein reductase (FabI), inhibited all clinical isolates of Staphylococcus aureus and Staphylococcus epidermidis at concentrations of ≤0.12 μg/ml.

Specifications

Chemical Name or Material AFN-1252
Physical Form Solid
CAS 620175-39-5
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C22H21N3O3
For Use With (Application) COVID-19-immunoregulation
Synonym API-1252 Debio 1452
Solubility Information DMSO : 5.8 mg/mL (15.45 mM; Need ultrasonic)
SMILES O=C(N(C)CC1=C(C)C2=CC=CC=C2O1)/C=C/C(C=C3CC4)=CNC3=NC4=O
Molecular Weight (g/mol) 375.42
Formula Weight 375.42
Percent Purity 99.13%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.