missing translation for 'onlineSavingsMsg'
Learn More

AFN-1252, MedChemExpress

Product Code. 16463227
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16463227 50 mg 50mg
16457208 10 mg 10mg
16449737 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16463227 Supplier Medchem Express Supplier No. HY16911
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress AFN-1252(Debio 1452) is a potent inhibitor of enoyl-acyl carrier protein reductase (FabI), inhibited all clinical isolates of Staphylococcus aureus and Staphylococcus epidermidis at concentrations of ≤0.12 μg/ml.

AFN-1252(Debio 1452) is a potent inhibitor of enoyl-acyl carrier protein reductase (FabI), inhibited all clinical isolates of Staphylococcus aureus and Staphylococcus epidermidis at concentrations of ≤0.12 μg/ml.

Specifications

Chemical Name or Material AFN-1252
Physical Form Solid
CAS 620175-39-5
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C22H21N3O3
For Use With (Application) COVID-19-immunoregulation
Synonym API-1252 Debio 1452
Solubility Information DMSO : 5.8 mg/mL (15.45 mM; Need ultrasonic)
SMILES O=C(N(C)CC1=C(C)C2=CC=CC=C2O1)/C=C/C(C=C3CC4)=CNC3=NC4=O
Molecular Weight (g/mol) 375.42
Formula Weight 375.42
Percent Purity 99.13%
Grade Research
Show More Show Less

Research purposes only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.