missing translation for 'onlineSavingsMsg'
Learn More

AFM Mouse anti-Human, Mouse, Clone: 2G9F6, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66113-1-IG-20UL

 View more versions of this product

Product Code. 16837695

  • £135.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This gene is a member of the albumin gene family, which is comprised of four genes that localize to chromosome 4 in a tandem arrangement. These four genes encode structurally-related serum transport proteins that are known to be evolutionarily related. The protein encoded by this gene is regulated developmentally, expressed in the liver and secreted into the bloodstream.
Specifications

Specifications

AFM
Monoclonal
0.55 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O89020, P43652
Afm
AFM Fusion Protein Ag16866
20 μL
Primary
Human, Mouse
Antibody
IgG1
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation, Western Blot
2G9F6
Unconjugated
AFM
afamin, afamin/AFM, AFM, ALB2, ALBA, ALF, Alpha Alb, Alpha albumin
Mouse
Protein G
RUO
173, 280662
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.