missing translation for 'onlineSavingsMsg'
Learn More

Aflatoxin B1, crystalline, Thermo Scientific Chemicals

Product Code. 10033150 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10033150 10 mg 10mg
10593011 1 mg 1mg
11966201 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10033150 Supplier Thermo Scientific Acros Supplier No. 227340100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Aflatoxin B1, crystalline, CAS # 1162-65-8, is a member of a group of mycotoxins presenting potent hepatotoxic and carcinogenic activities.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Information

• Aflatoxin B1 (AFB1) is a member of a group of mycotoxins produced by Aspergillus flavus, and Aspergillus parasiticus showing carcinogenic activities
• AFB1 can induce the transversion of G to T in the third position of codon 249 of the p53 tumor suppressor gene, resulting in a mutation and promoting the carcinogenic process

Applications

• Aflatoxin B1 shows teratogenic activity
• It is a CYP1A2, CYP2A6, CYP2D6, and CYP3A substrate and can be used to study the CYP family in in vitro and in vivo studies.
• It can cause immunosuppression in animal models.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1162-65-8
Molecular Formula C17H12O6
Molecular Weight (g/mol) 312.28
MDL Number MFCD00869647
InChI Key OQIQSTLJSLGHID-UHFFFAOYNA-N
Synonym aflatoxin b1, ccris 12, unii-9n2n2y55mh, afb1, 2,3,6aalpha,9aalpha-tetrahydro-4-methoxycyclopenta c furo 3',2':4,5 furo 2,3-h 1 benzopyran-1,11-dione, 1h,11h-cyclopenta c furo 3',2':4,5 furo 2,3-h 1 benzopyran-1,11-dione,2,3,6a,9a-tetrahydro-4-methoxy-, 6ar,9as, cyclopenta c furo 3',2':4,5 furo 2,3-h 1 benzopyran-1,11-dione, 2,3,6a,9a-tetrahydro-4-methoxy, cyclopenta c furo 3',2':4,5 furo 2,3-h 1 benzopyran-1,11-dione, 2,3,6a,9a-tetrahydro-4-methoxy-, 6ar,9as, aflatoxin-b1, aflatoxin b
PubChem CID 186907
ChEBI CHEBI:2504
IUPAC Name 11-methoxy-6,8,19-trioxapentacyclo[10.7.0.0²,⁹.0³,⁷.0¹³,¹⁷]nonadeca-1,4,9,11,13(17)-pentaene-16,18-dione
SMILES COC1=C2C3=C(C(=O)CC3)C(=O)OC2=C2C3C=COC3OC2=C1

Specifications

Packaging Glass Bottle
Quantity 10 mg
Merck Index 15, 171
Formula Weight 312.27
Chemical Name or Material Aflatoxin B1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.