missing translation for 'onlineSavingsMsg'
Learn More

Aflatoxin B1 Antibody (6A10), mFluor Violet 500 SE, Novus Biologicals™

Product Code. 18414304
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18414304 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18414304 Supplier Novus Biologicals (Bio-Techne) Supplier No. NB600443MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Aflatoxin B1 Monoclonal antibody specifically detects Aflatoxin B1 in Human, All samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Dot Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aflatoxin B1
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 6A10
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Dot Blot
Formulation 50mM Sodium Borate
Gene Alias AFB1, AFB1-AR1, Aldoketoreductase 7
Host Species Mouse
Immunogen Aflatoxin B1 DNA (against the midazole ring-opened persistent form of the major N-7 guanine adduct of AFB1).
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, All species
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.