missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aflatoxin B1 Antibody (6A10), mFluor Violet 450 SE, Novus Biologicals™
Description
Aflatoxin B1 Monoclonal antibody specifically detects Aflatoxin B1 in Human, All samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Dot Blot
Specifications
Specifications
| Antigen | Aflatoxin B1 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 6A10 |
| Conjugate | mFluor Violet 450 SE |
| Dilution | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Dot Blot |
| Formulation | 50mM Sodium Borate |
| Gene Alias | AFB1, AFB1-AR1, Aldoketoreductase 7 |
| Host Species | Mouse |
| Immunogen | Aflatoxin B1 DNA (against the midazole ring-opened persistent form of the major N-7 guanine adduct of AFB1). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?