missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aflatoxin B1 Antibody (6A10), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB600-443AF405
This item is not returnable.
View return policy
Description
Aflatoxin B1 Monoclonal specifically detects Aflatoxin B1 in Human, All Species samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| Aflatoxin B1 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| AFB1, AFB1-AR1, Aldoketoreductase 7 | |
| Aflatoxin B1 DNA (against the midazole ring-opened persistent form of the major N-7 guanine adduct of AFB1). | |
| 0.1 mL | |
| Primary | |
| Human, All species | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| 6A10 | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| Mouse | |
| Protein G purified | |
| RUO | |
| All species. There is no cross-reactivity with other carcinogen-DNA adducts. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction