missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Shrimp Alkaline Phosphatase (SAP)

Product Code. 15523687
Change view
Click to view available options
Quantity:
1000 U
500 U
5000 U
Unit Size:
1000 units
500 units
5000 units
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15523687 1000 U 1000 units
15533687 500 U 500 units
15543687 5000 U 5000 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15523687 Supplier Invitrogen™ Supplier No. 783901000UN
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Treat unincorporated dNTPs in PCR reactions to prepare templates for DNA sequencing or SNP analysis. Shrimp Alkaline Phosphatase (SAP) is useful in molecular biology applications such as the dephosphorylation of phosphorylated ends of DNA or RNA for subsequent use in cloning or end-labeling of probes.

  • 100% heat-inactivated in 5 min at 65°C
  • Significantly improved storage stability at lower temperatures
  • Very high specific activity (see Fig. 3)
  • Removes 5’-phosphates from DNA, RNA, dNTPs, and proteins
  • Purified from a recombinant source
  • May be added directly to restriction enzyme digests
  • No vector purification necessary
  • Requires no supplemental zinc or other additives for activity
  • Works direct in many different buffers
  • Easy treatment of unincorporated dNTPs in PCR products prior to DNA sequencing or SNP analysis
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Dephosphorylation of DNA/RNA
Quantity 1000 U
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.