missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Exonuclease I

Product Code. 15513677
Change view
Click to view available options
Quantity:
2500 U
Unit Size:
2500 units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15513677 2500 U 2500 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15513677 Supplier Invitrogen™ Supplier No. 70073Z2500UN

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Useful in preparing PCR products for applications involving sequencing or labeling methods

Exonuclease I is particularly useful in preparing the products of PCR for applications involving sequencing or labeling methods1. Typically, the excess primers and any other extraneous single-stranded DNA present in PCR products will interfere with subsequent enzymatic reactions involving DNA synthesis. The hydrolytic properties of Exonuclease I degrade all single-stranded DNA present in the PCR mixture allowing the product to be used more efficiently in other applications.

  • Hydrolyzes single-stranded DNA in the 3′-5′ direction, releasing 5′-mononucleotides and leaving the terminal 5′-dinucleotide intact; hydrolysis is processive and cannot proceed if the 3′ terminus is phosphorylated
  • Use to measure the endonucleolytic cleavage of covalently closed circular single-stranded DNA reacted with an endonuclease of interest2
  • Also suitable for measuring DNA helicase activity3
  • When combined with Shrimp Alkaline Phosphatase (Mfr. No. 78390) for dNTP dephosphorylation, the use of alternative purification methods, such as columns, gels or magnetic separations, are completely eliminated

Source: E. coli strain containing an overproducing clone of E. coli Exonuclease I
Properties:

  • Molecular Weight: 55 kDa
  • Heat Inactivation: 80° for 15 min.
  • Degrades to terminal dinucleotides
  • Degrades glycosylated DNA
  • Optimum Temperature: 37°C
Purity:
  • Greater than 95% pure as determined by SDS-PAGE
  • Tested for contaminating endonucleases, double-stranded exonucleases and ribonucleases
Storage Buffer:
20mM Tris-HCI (pH 7.5), 5mM 2-mercaptoethanol, 0.5mM EDTA, 50% glycerol
Assay Conditions:
  • Reaction mixture (100μL) contains 67mM glycine buffer (pH 9.5), 10mM
    2-mercaptoethanol, 6.7mM MgCl2, 0.5mM denatured DNA and enzyme
  • Incubation is at 37°C for 30 min.
  • Unit Definition: One unit is the amount of enzyme which catalyzes the release of 10nmol of acid-soluble nucleotide from denatured DNA in 30 min at 37°C under standard conditions
    Concentration:
    • Standard Conc.: 10 units/μL, Mfr. No. 70073
    • High Conc.: 20 units/μL, Mfr. No. 72073
    Functional Assay: Treated PCR product with Exonuclease I to degrade unincorporated primers before performing sequencing reaction with Sequenase™ Version 2.0 DNA Polymerase Sequencing Kit (Mfr. No. 70770)

    Elimination of residual single-stranded DNA containing a 3′ terminus; Measuring endonucleolytic cleavage of covalently closed circular ssDNA; Measuring DNA helicase activity

    TRUSTED_SUSTAINABILITY

    Specifications

    Concentration 10 U/μL
    Quantity 2500 U
    Product Type Exonuclease I
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.