missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
AffiniPure Polyclonal antibody specifically detects AffiniPure in Goat samples. It is validated for ELISA, Western Blot.
Specifications
Specifications
| Antigen | AffiniPure |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | HRP |
| Host Species | Rabbit |
| Quantity | 200 μL |
| Primary or Secondary | Secondary |
| Target Species | Goat |
| Form | Liquid |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?