missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AFF4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13197-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene belongs to the AF4 family of transcription factors involved in leukemia. It is a component of the positive transcription elongation factor b (P-TEFb) complex. A chromosomal translocation involving this gene and MLL gene on chromosome 11 is found in infant acute lymphoblastic leukemia with ins.Specifications
| AFF4 | |
| Polyclonal | |
| Unconjugated | |
| AFF4 | |
| AF4/FMR2 family member 4, AF4/FMR2 family, member 4, AF5Q31, AFF4, MCEF, Protein AF 5q31 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 27125, 303132, 93736 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9ESC8, Q9UHB7 | |
| Aff4 | |
| AFF4 Fusion Protein Ag3931 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction