missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AFF4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554866
This item is not returnable.
View return policy
Description
Immunogen sequence: QGTGNSYTDT SGPKETSSAT PGRDSKTIQK GSESGRGRQK SPAQSDSTTQ RRTVGKKQPK KAEKAAAEEP RGGLKIESET PVDLASSMPS SRHKAATKGS RKPNIKKESK SSPRPTAEKK KYKSTSKSSQ KS Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.
The protein encoded by this gene belongs to the AF4 family of transcription factors involved in leukemia. It is a component of the positive transcription elongation factor b (P-TEFb) complex. A chromosomal translocation involving this gene and MLL gene on chromosome 11 is found in infant acute lymphoblastic leukemia with ins.
Specifications
| AFF4 | |
| Polyclonal | |
| Unconjugated | |
| AFF4 | |
| AF4/FMR2 family member 4; AF4/FMR2 family, member 4; AF5Q31; Aff4; Alf4; ALL1 fused gene from 5q31; ALL1-fused gene from chromosome 5q31 protein; CHOPS; HSPC092; Laf4l; LOW QUALITY PROTEIN: AF4/FMR2 family member 4; major CDK9 elongation factor-associated protein; MCEF; Protein AF-5q31; RA_m002_jsmFBA6Br | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 27125 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UHB7 | |
| AFF4 | |
| Recombinant protein corresponding to Human AFF4. Recombinant protein control fragment (Product #RP-89295). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction