missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AFF4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-40357
This item is not returnable.
View return policy
Description
AFF4 Polyclonal specifically detects AFF4 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative).
Specifications
| AFF4 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| AFF4 | |
| The immunogen for this product maps to a region between residue 1113 and 1163 of human major CDK9 elongation factor-associated protein of human using the numbering given in entry NP_055238.1 (GeneID 27125). | |
| 100 μL | |
| Primary | |
| Human | |
| IgG |
| Immunoprecipitation, Western Blot | |
| 1.0 mg/mL | |
| Immunoprecipitation 5-15 ug/mg lysate, Western Blot (Negative) | |
| AF4/FMR2 family, member 4, AF5Q31AF4/FMR2 family member 4, ALL1-fused gene from chromosome 5q31 protein, Major CDK9 elongation factor-associated protein, MCEFALL1 fused gene from 5q31, MGC75036, Protein AF-5q31 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 27125 | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction