missing translation for 'onlineSavingsMsg'
Learn More

AFF4 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18408752 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18408752 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18408752 Supplier Novus Biologicals Supplier No. NBP140356JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AFF4 Polyclonal antibody specifically detects AFF4 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen AFF4
Applications Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 646
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias AF4/FMR2 family, member 4, AF5Q31AF4/FMR2 family member 4, ALL1-fused gene from chromosome 5q31 protein, Major CDK9 elongation factor-associated protein, MCEFALL1 fused gene from 5q31, MGC75036, Protein AF-5q31
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 50 and 100 of human major CDK9 elongation factor-associated protein of human using the numbering given in entry NP_055238.1 (GeneID 27125).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27125
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.