missing translation for 'onlineSavingsMsg'
Learn More

Afegostat D-Tartrate, MedChemExpress

Product Code. 16443446
Encompass_Preferred
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 16443446

Brand: Medchem Express HY14829E

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Afegostat D-Tartrate is a pharmacological chaperone, which specifically and reversibly binds acid-β-glucosidase (GCase) in the endoplasmic reticulum (ER) with high affinity.

Afegostat D-Tartrate is a pharmacological chaperone, which specifically and reversibly binds acid-β-glucosidase (GCase) in the endoplasmic reticulum (ER) with high affinity.

Specifications

Chemical Name or Material Afegostat D-Tartrate
Physical Form Solid
CAS 957230-65-8
Color Yellow
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C10H19NO9
For Use With (Application) Neuroscience-Neurodegeneration
Synonym D-Isofagomine D-TartrateIsofagomine D-Tartrate
Solubility Information H2O : ≥ 50 mg/mL (168.20 mM)
SMILES OC[C@@H]1[C@H]([C@@H](CNC1)O)O.OC([C@@H](O)[C@H](O)C(O)=O)=O
Molecular Weight (g/mol) 297.26
Formula Weight 297.26
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.