missing translation for 'onlineSavingsMsg'
Learn More

Afatinib, MedChemExpress

Product Code. 16434816
Encompass_Preferred
missing translation for 'orderingAttributeHoverText'
Quantity:
10 mg
50 mg
100 mg
200 mg
missing translation for 'unitSize'
100mg
10mg
200mg
50mg
This item is not returnable. View return policy

Product Code. 16434816

missing translation for 'mfr': Medchem Express HY10261

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Afatinib (BIBW 2992) is an orally active, potent and irreversible dual specificity inhibitor of ErbB family (EGFR and HER2), with IC50 values of 0.5 nM, 0.4 nM, 10 nM and 14 nM for EGFRwt, EGFRL858R, EGFRL858R/T790M and HER2, respectively. Afatinib can be used for the research of esophageal squamous cell carcinoma (ESCC), non-small cell lung cancer (NSCLC) and gastric cancer.

Afatinib (BIBW 2992) is an orally active, potent and irreversible dual specificity inhibitor of ErbB family (EGFR and HER2), with IC50 values of 0.5 nM, 0.4 nM, 10 nM and 14 nM for EGFRwt, EGFRL858R, EGFRL858R/T790M and HER2, respectively. Afatinib can be used for the research of esophageal squamous cell carcinoma (ESCC), non-small cell lung cancer (NSCLC) and gastric cancer.

Specifications

Chemical Name or Material Afatinib
CAS 850140-72-6
Color White
Purity Grade Notes Research
Molecular Formula C24H25ClFN5O3
Quantity 100 mg
Synonym BIBW 2992
Solubility Information DMSO : 100 mg/mL (205.79 mM; Need ultrasonic)
SMILES O=C(NC1=C(C=C2C(C(NC3=CC(Cl)=C(C=C3)F)=NC=N2)=C1)O[C@H]4CCOC4)/C=C/CN(C)C.[(E)]
Molecular Weight (g/mol) 485.94
Formula Weight 485.94
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.