missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Afatinib, MedChemExpress
Description
Afatinib (BIBW 2992) is an orally active, potent and irreversible dual specificity inhibitor of ErbB family (EGFR and HER2), with IC50 values of 0.5 nM, 0.4 nM, 10 nM and 14 nM for EGFRwt, EGFRL858R, EGFRL858R/T790M and HER2, respectively. Afatinib can be used for the research of esophageal squamous cell carcinoma (ESCC), non-small cell lung cancer (NSCLC) and gastric cancer.
Specifications
Specifications
| Chemical Name or Material | Afatinib |
| CAS | 850140-72-6 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C24H25ClFN5O3 |
| Quantity | 10 mg |
| Synonym | BIBW 2992 |
| Solubility Information | DMSO : 100 mg/mL (205.79 mM; Need ultrasonic) |
| SMILES | O=C(NC1=C(C=C2C(C(NC3=CC(Cl)=C(C=C3)F)=NC=N2)=C1)O[C@H]4CCOC4)/C=C/CN(C)C.[(E)] |
| Molecular Weight (g/mol) | 485.94 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?