missing translation for 'onlineSavingsMsg'
Learn More

Afatinib-des(4-dimethylamino-2-en-1-oxo)butyl (Contained ~8.5% Ethanol), TRC

Product Code. 30010497
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010497 1 g 1g
30008026 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30010497 Supplier Toronto Research Chemicals Supplier No. TRCA3553201G
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 314771-76-1
Molecular Formula C18H16ClFN4O2
Molecular Weight (g/mol) 374.8
Synonym N4-(3-Chloro-4-fluorophenyl)-7-[[(3S)-tetrahydro-3-furanyl]oxy]-4,6-quinazolinediamine, (S)-N4-(3-Chloro-4-fluorophenyl)-7-((tetrahydrofuran-3-yl)oxy)quinazoline-4,6-diamine, Des(4-dimethylamino-2-en-1-oxo)butyl Afatinib
IUPAC Name 4-N-(3-chloro-4-fluorophenyl)-7-[(3S)-oxolan-3-yl]oxyquinazoline-4,6-diamine
SMILES ClC1=C(F)C=CC(NC2=NC=NC3=CC(O[C@H]4CCOC4)=C(N)C=C32)=C1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C18H16ClFN4O2/c19-13-5-10(1-2-14(13)20)24-18-12-6-15(21)17(7-16(12)22-9-23-18)26-11-3-4-25-8-11/h1-2,5-7,9,11H,3-4,8,21H2,(H,22,23,24)/t11-/m0/s1
Chemical Name or Material Afatinib-des(4-dimethylamino-2-en-1-oxo)butyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.