missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
AFAP1L2 Polyclonal specifically detects AFAP1L2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
| Antigen | AFAP1L2 |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | FITC |
| Dilution | Immunocytochemistry/Immunofluorescence |
| Formulation | PBS |
| Gene Alias | actin filament associated protein 1-like 2, actin filament-associated protein 1-like 2, AFAP1-like protein 2, FLJ14564, KIAA1914Em:AC005383.4, XB130CTB-1144G6.4 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human AFAP1L2 fragment. |
| Purification Method | Antigen and protein A Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?