missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AFAP1L1 Rabbit anti-Human, Non-human primate, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23909-1-AP-20UL
This item is not returnable.
View return policy
Description
May be involved in podosome and invadosome formation.Specifications
| AFAP1L1 | |
| Polyclonal | |
| Unconjugated | |
| AFAP1L1 | |
| AFAP1 like protein 1, AFAP1L1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 134265 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.34 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8TED9 | |
| AFAP1L1 | |
| AFAP1L1 Fusion Protein Ag21006 | |
| 20 μL | |
| Primary | |
| Non-human Primate, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction