Learn More
AFAP1L1 Rabbit anti-Human, Non-human primate, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 23909-1-AP-150UL
 View more versions of this product
Product Code. 16872515
  • £349.00 / 150µL
Description
May be involved in podosome and invadosome formation.
Specifications
Specifications
AFAP1L1
Polyclonal
Unconjugated
AFAP1L1
AFAP1 like protein 1, AFAP1L1
Rabbit
Antigen Affinity Chromatography
RUO
134265
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.34 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8TED9
AFAP1L1
AFAP1L1 Fusion Protein Ag21006
150 μL
Primary
Human, Non-human Primate