missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AFAP Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA547636
This item is not returnable.
View return policy
Description
This antibody detects endogenous human AFAP in Western blots. Reconstitute at 0.2 mg/mL in sterile PBS.
The protein encoded by this gene is a Src binding partner. It may represent a potential modulator of actin filament integrity in response to cellular signals, and may function as an adaptor protein by linking Src family members and/or other signaling proteins to actin filaments. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| AFAP | |
| Polyclonal | |
| Unconjugated | |
| Afap1 | |
| 110 kDa actin filament-associated protein; 2600003E23Rik; 9630044L16Rik; actin filament associated protein 1; actin filament-associated protein 1; actin filament-associated protein, 110 kDa; AFAP; Afap1; AFAP110; AFAP-110; AI848729; FLJ56849; im:7148276; Kiaa3018; mKIAA3018 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 60312 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q8N556 | |
| Afap1 | |
| E. coli-derived recombinant human AFAP His450-Thr730. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction