missing translation for 'onlineSavingsMsg'
Learn More

AFAP Antibody (OTI2G9), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-72120PCP

Product Code. 30030020

  • £561.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

AFAP Monoclonal antibody specifically detects AFAP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

AFAP
Monoclonal
PerCP
PBS
Mouse
Immunogen affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot, Immunohistochemistry
OTI2G9
Western Blot, Immunohistochemistry
actin filament associated protein 1, FLJ56849
Human recombinant protein fragment corresponding to amino acids 100-386 of human AFAP1 (NP_940997) produced in E.coli.
0.1 mL
Cancer, Cell Cycle and Replication
60312
Store at 4C in the dark.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.