missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Afamin Monoclonal Antibody (C1), Invitrogen™

Mouse Monoclonal Antibody
£161.00 - £389.00
Specifications
| Antigen | Afamin |
|---|---|
| Clone | C1 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17970073
|
Invitrogen™
MA551042 |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17900153
![]() |
Invitrogen™
MA551041 |
200 μL |
£389.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene is a member of the albumin gene family, which is comprised of four genes that localize to chromosome 4 in a tandem arrangement. These four genes encode structurally-related serum transport proteins that are known to be evolutionarily related. The protein encoded by this gene is regulated developmentally, expressed in the liver and secreted into the bloodstream.Specifications
| Afamin | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| Afamin; AFM; ALB2; ALBA; Alf; alpha albumin; alpha-Alb; Alpha-albumin; MGC125338; MGC125339 | |
| AFM | |
| IgG1 κ | |
| Protein A/G | |
| AFM |
| C1 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Rat | |
| P36953 | |
| 282708 | |
| Recombinant Rat AFM protein, Leu22-Tyr201 (Accession #P36953). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title